Learn More
Sino Biological Human TEK PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10700RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human TIE2 (rh TIE2; Catalog#10700-H08H; AAA61139.1; Met 1-Lys 745). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-1354