Learn More
ACROBiosystems Human TEM8/ANTXR1 200ug
Catalog No. 502534069
Encompass_Preferred
Biotinylated Human TEM8 / ANTXR1 Protein HisAvitag (TE8-H82E3) is expressed from human 293 cells (HEK293). It contains AA Glu 33 - Ser 321 (Accession # Q9H6X2).
Catalog No. 50-253-4069 Supplier ACROBiosystems Supplier No. TE8H82E3200UG
Biotinylated Human TEM8 / ANTXR1 Protein HisAvitag (TE8-H82E3) is expressed from human 293 cells (HEK293). It contains AA Glu 33 - Ser 321 (Accession # Q9H6X2).