Learn More
Raybiotech Inc Human Tenascin C ELISA Kit
Supplier:  Raybiotech Inc ELHTNC1
Encompass_Preferred
For serum, plasma, and cell culture supernatant samples. Sensitivity: 6.1 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human Tenascin C. Other species not determined.
Catalog No. 50-194-9790