Learn More
Biomatik Corporation Human Tensin 1 (TNS1) ELISA Kit
Supplier:  Biomatik Corporation EKU12344
Encompass_Preferred
Human Tensin 1 (TNS1) ELISA Kit for detecting TNS1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.127ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9HBL0. Gene ID: 7145. Target Synonyms: TNS; MXRA6; Matrix-Remodelling Associated 6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5523