Learn More
Biomatik Corporation Human Teratocarcinoma Derived Growth Factor 1 (TDGF1) CLIA Kit
Supplier:  Biomatik Corporation EKU09582
Encompass_Preferred
Human Teratocarcinoma Derived Growth Factor 1 (TDGF1) CLIA Kit for detecting TDGF1 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 13.7-10,000pg/ml. Sensitivity: 5.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P13385. Gene ID: 6997. Target Synonyms: CR; CRGF; CRIPTO; Cripto-1; Epidermal growth factor-like cripto protein CR1; Cripto-1 growth factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8491