Learn More
Biomatik Corporation Human TERF1 Interacting Nuclear Factor 2 (TINF2) ELISA Kit
Supplier:  Biomatik Corporation EKU07598
Encompass_Preferred
Human TERF1 Interacting Nuclear Factor 2 (TINF2) ELISA Kit for detecting TINF2 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.068ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9BSI4. Gene ID: 26277. Target Synonyms: TIN2; TRF1-interacting nuclear protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7381