Learn More
Biomatik Corporation Human Testis Anion Transporter 1 (TAT1) ELISA Kit
Supplier:  Biomatik Corporation EKU07604
Encompass_Preferred
Human Testis Anion Transporter 1 (TAT1) ELISA Kit for detecting TAT1 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.118ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q96RN1. Gene ID: 116369. Target Synonyms: SLC26A8; Solute Carrier Family 26 Member 8; Anion exchange transporter. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6760