Learn More
Sino Biological Anti-Transferrin Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11019MM0620
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Transferrin / TF (rh Transferrin; Catalog#11019-H08H; NP_001054.1; Met 1-Pro 698). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-0337