Learn More
BIOHIPPO Human TF Microsample
SDP
Catalog No. 5000313963
Encompass
Human TF Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TF in plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 6 25-400 pg/mL with a sensitivity of 2 67 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TF Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TF Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TF biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13963 Supplier BIOHIPPO Supplier No. BHE15210537
Human TF Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TF in plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 6 25-400 pg/mL with a sensitivity of 2 67 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TF Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TF Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TF biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color