Learn More
Sino Biological Anti-DP2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101770T10100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human DP2, and purified by antigen affinity chromatography.
Catalog No. 50-241-1394