Learn More
Raybiotech Inc Human TFF3 (Trefoil Factor 3) ELISA Kit
Catalog No. 501949765
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 1.5 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human TFF-3. Other species not determined.
Catalog No. 50-194-9765 Supplier Raybiotech Inc Supplier No. ELHTFF31
Add to Cart
Add to Cart
For cell culture supernatants, plasma, and serum samples. Sensitivity: 1.5 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human TFF-3. Other species not determined.