Learn More
Sino Biological Recombinant Human TFF1/Trefoil Factor 1 Protein (His Tag)
Supplier:  Sino Biological 13629H08H100
Encompass_Preferred
A DNA sequence encoding the human TFF1 (NP_003216.1) (Met1-Phe84) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-4208