Learn More
Sino Biological Human TFPI Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000786546
Encompass_Preferred
The Human TFPI ELISA Pair Set is for the quantitative determination of Human TFPI.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human TFPI coated on a 96-well plate. Standards and samples are added to the wells, and any Human TFPI present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human TFPI monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human TFPI present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86546 Supplier Sino Biological Supplier No. SEK105645
The Human TFPI ELISA Pair Set is for the quantitative determination of Human TFPI.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human TFPI coated on a 96-well plate. Standards and samples are added to the wells, and any Human TFPI present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human TFPI monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human TFPI present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.