Learn More
Sino Biological Anti-TFPI2 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10545MM04100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TFPI2 (rh TFPI2; Catalog#10545-H08H; NP_006519.1; Met 1-Lys 213). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-1601