Learn More
Sino Biological Recombinant Anti-Transferrin Receptor/TFR1/CD71 Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 11020R040A100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Transferrin Receptor / TFRC / CD71 (rh Transferrin Receptor / TFRC / CD71; Catalog#11020-H07H; AAA61153.1; Cys89-Phe760) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-241-2323