Learn More
Sino Biological Recombinant Anti-Transferrin Receptor/TFR1/CD71 Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 11020R040F25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Transferrin Receptor / TFRC / CD71 (rh Transferrin Receptor / TFRC / CD71; Catalog#11020-H07H; AAA61153.1; Cys89-Phe 760) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-241-1170