Learn More
Sino Biological Anti-Transferrin Receptor/TFR1/CD71 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 11020MM03F25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Transferrin Receptor / TFRC / CD71 (rh Transferrin Receptor / TFRC / CD71; Catalog#11020-H07H; AAA61153.1; Cys89-Phe760) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-241-1888