Learn More
Sino Biological Human TFRC MAb 100ul (Sino Biological)
Supplier:  Sino Biological 11020R001100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Transferrin Receptor / TFRC / CD71 (rh Transferrin Receptor / TFRC / CD71; Catalog#11020-H07H; AAA61153.1; Cys89-Phe 760).
Catalog No. 50-237-2387