Learn More
BIOHIPPO Human TG Microsample
SDP
Catalog No. 5000315551
Encompass
Human TG Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TG in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 136 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TG Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TG Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TG biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15551 Supplier BIOHIPPO Supplier No. BHE15212038
Human TG Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TG in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 136 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TG Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TG Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TG biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color