Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TG native Protein from human thyroid

Catalog No. 89964522 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-964-522 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-964-522 Supplier Abnova™ Supplier No. P5343
Add to Cart
Add to Cart

Used for SDS-PAGE

Specifications

For Use With (Application) SDS-PAGE
Formulation Lyophilized from 0.02M ammonium bicarbonate andMay contain traces of buffer salts
Gene ID (Entrez) 7038
Name TG (Human) Native Protein
Preparation Method Native protein purified from human thyroid
Quantity 5 mg
Storage Requirements Store at 4°C. This product is stable for at least 3 months. After reconstitution with phosphate buffer, pH 7.0 (contains 0.15 M NaCl), aliquot and store at 4°C.
Regulatory Status RUO
Gene Alias AITD3/TGN
Common Name TG
Gene Symbol TG
Species Human
Protein Tag None
Expression System Native protein purified from human thyroid
Form Lyophilized
Purity or Quality Grade ≥96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.