Learn More
Bon Opus Biosciences Recombinant Human Transforming Growth Factor β-1/TGFB1
SDP
Supplier:  Bon Opus Biosciences CA5910UG
Encompass_Preferred
Recombinant Human Transforming Growth Factor beta 1 is produced by our Mammalian expression system and the target gene encoding Ala279-Ser390 is expressed. Bon Opus Cat. #CA59
Catalog No. 50-725-143