Learn More
Raybiotech Inc Human TGF alpha ELISA Kit
Supplier:  Raybiotech Inc ELHTGFA1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 3 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA kit shows no cross-reactivity with any of the cytokines tested: Human ALCAM, Angiopoietin-1, Angiostatin, B7-1, BMP-7, CD30, CD40, CD40 Ligand, CTLA-4, CXCL16, Dkk-4 , DR6, Endostatin, E-Selectin, HB-EGF, ICAM-2, IGF-II, IL-10 Ra, IL-10 Rb, IL-13 Ra1, IL-18 BPa, IL-18 Rb, IL-18, IL-2 Ra, MMP-1, IL-2 Rb, IL-2 Rg, IL-21 R, MMP-2 , MMP-3, MMP-10, MMP-13).
Catalog No. 50-194-9768