Learn More
TRIBIOSCIENCE INC Human TGF-β1 ELISA, 1 x 96-well plate
SDP
Catalog No. NC3454589
Encompass_Preferred
Tribioscience’s Human TGF-β1 Fast ELISA is designed to quantitatively detect human TGF-β1 levels in in CSF, brain tissue, serum, plasma, and other biological samples. The kit uses proprietary methods to combine samples and detections in one step, rather than complex traditional methods. The detection range is from 10 to 4,000 pg/mL.
Catalog No. NC3454589 Supplier TRIBIOSCIENCE INC Supplier No. TBS3232
Tribioscience’s Human TGF-β1 Fast ELISA is designed to quantitatively detect human TGF-β1 levels in in CSF, brain tissue, serum, plasma, and other biological samples. The kit uses proprietary methods to combine samples and detections in one step, rather than complex traditional methods. The detection range is from 10 to 4,000 pg/mL.