Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human/Mouse/Rat/Porcine/Canine TGF-beta 1 Quantikine ELISA

Catalog No. SB100B
Change view
Click to view available options
Quantity:
6 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB100B 6 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SB100B Supplier R&D Systems™ Supplier No. SB100B
This item is not returnable. View return policy

Human TGF-beta 1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human TGF-beta 1 Quantikine ELISA Kit quantifies human TGF-beta 1 in cell culture supernates (100 ul), serum (40 ul), edta plasma (40 ul), urine (100 ul).

Specifications

Assay Range 31.20 - 2000pg/mL
Assay Sensitivity 15.4pg/mL
Conjugate HRP
Cross Reactivity Cross-species reactivity observed with 1 or more species tested. Cross-reactivity observed with 1 or more available related molecules
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA), Platelet-poor Plasma, Serum, Urine
Sufficient For 6 x 96-well Plates
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 7040
Quantity 6 Plates
Sample Volume Cell Culture Supernates (100uL),Serum (40uL),EDTA Plasma (40uL),Urine (100uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.