Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Human TGF-Beta 1 (TGFB1) Recombinant Protein, HEK293-expressed, Lyophilized, 1 mg (10 x 100 μg)

Catalog No. NC3357818
Encompass_Preferred

Recombinant human transforming growth factor-beta 1 (TGF-β1) expressed in HEK293 cells and supplied lyophilized in a trehalose-stabilized formulation. Contains amino acids Ala 279-Ser 390 (calculated MW 12.8 kDa), >95% purity, and low endotoxin; validated for cell-based bioactivity and ELISA applications.

  • Human sequence containing amino acids Ala 279-Ser 390.
  • HEK293-expressed, tag-free recombinant protein.
  • Lyophilized formulation with trehalose protectant.
  • Purity greater than 95% by SDS-PAGE.
  • Specific activity >6.00 x 10^6 IU/mg (bioassay calibrated to NIBSC 89/514).
  • Endotoxin less than 0.01 EU per μg.
  • Host cell protein and DNA minimized (HCP <0.5 ng/μg; HCD <0.02 ng/μg).
  • Vial options include 20 μg, 50 μg, and 1 mg (100 μg x 10).
  • Storage: stable lyophilized at -20°C to -70°C; store reconstituted material at -70°C.

Catalog No. NC3357818 Supplier ACROBiosystems Supplier No. TG1H52181MG10X100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human transforming growth factor-beta 1 (TGF-β1) expressed in HEK293 cells and supplied lyophilized in a trehalose-stabilized formulation. Contains amino acids Ala 279-Ser 390 (calculated MW 12.8 kDa), >95% purity, and low endotoxin; validated for cell-based bioactivity and ELISA applications.

  • Human sequence containing amino acids Ala 279-Ser 390.
  • HEK293-expressed, tag-free recombinant protein.
  • Lyophilized formulation with trehalose protectant.
  • Purity greater than 95% by SDS-PAGE.
  • Specific activity >6.00 x 10^6 IU/mg (bioassay calibrated to NIBSC 89/514).
  • Endotoxin less than 0.01 EU per μg.
  • Host cell protein and DNA minimized (HCP <0.5 ng/μg; HCD <0.02 ng/μg).
  • Vial options include 20 μg, 50 μg, and 1 mg (100 μg x 10).
  • Storage: stable lyophilized at -20°C to -70°C; store reconstituted material at -70°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.