Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human TGF-beta 2 Quantikine ELISA Kit

Catalog No. DB250
Change view
Click to view available options
Quantity:
1 Plate
6 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DB250 1 Plate
SB250 6 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. DB250 Supplier R&D Systems™ Supplier No. DB250
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human TGF-beta 2 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human TGF-beta 2 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human TGF-beta 2 in cell culture supernates (125 ul), serum (125 ul), edta plasma (125 ul), heparin plasma (125 ul), citrate plasma (125 ul).

Specifications

Assay Range 31.20 - 2000pg/mL
Assay Sensitivity 7pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
Specificity Natural and Recombinant Human TGF-beta 2 and natural Porcine TGF-beta 2.
Sufficient For 1 x 96-well Plate
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 7042
Quantity 1 Plate
Sample Volume Cell Culture Supernates (125uL),Serum (125uL),EDTA Plasma (125uL),Heparin Plasma (125uL),Citrate Plasma (125uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym BSC-1 cell growth inhibitor, Cetermin, Glioblastoma-derived T-cell suppressor factor, G-TSF, MGC116892, Polyergin, TGFB2, TGFbeta 2, TGF-beta2, TGF-beta-2, transforming growth factor beta-2, transforming growth factor, beta 2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.