Learn More
Gibco™ Human TGF-beta 2 Recombinant Protein, PeproTech®
Description
100-35B-1MG will be provided as 4 x 250 μg (100-35B-250UG). 100-35B-500UG will be provided as 2 x 250 μg (100-35B-250UG). Recombinant Human TGF-beta2 is a 25.0 kDa protein composed of two identical 112 amino acid polypeptide chains linked by a single disulfide bond. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis.
Specifications
Specifications
| Accession Number | P61812 |
| For Use With (Application) | Control, Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 7042 |
| Molecular Weight (g/mol) | 25 kDa |
| Name | Human TGF-beta 2 |
| Purification Method | Purified |
| Quantity | 2 μg |
| Source | HEK293 cells |
| Storage Requirements | -20°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.