Learn More
ACROBiosystems ACROBiosystems Human TGF-Beta 2 / TGFB2 Protein, Tag Free
Catalog No. 502107080
Encompass_Preferred
Recombinant Protein;10UG;Human TGF-Beta 2, Tag Free (TG2-H4215) is expressed from human 293 cells (HEK293). It contains AA Ala 303 - Ser 414 (Accession # P61812-1).;This protein carries no "tag". The protein has a calculated MW of 12.7 kDa. The protein migrates as 13-14 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-7080 Supplier ACROBiosystems Supplier No. TG2H421510UG
Recombinant Protein;10UG;Human TGF-Beta 2, Tag Free (TG2-H4215) is expressed from human 293 cells (HEK293). It contains AA Ala 303 - Ser 414 (Accession # P61812-1).;This protein carries no "tag". The protein has a calculated MW of 12.7 kDa. The protein migrates as 13-14 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.