Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human TGF beta-3 ELISA Kit

Catalog No. EH613RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH613RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EH613RB Supplier Invitrogen™ Supplier No. EH613RB
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Human TGF beta-3 ELISA Kit is an enzyme-linked immunosorbent assay for quantitative detection of human TGF beta 3 in cell culture supernatants and serum samples.

Transforming growth factor beta 3 (TGF beta 3) is the third member of the transforming growth factor family of cytokines, which also includes TGF beta 1 and beta 2. These cytokines are secreted in precursor form consisting of a bioactive C-terminal domain attached to an N-terminal domain known as latency associated protein (LAP). Cleavage of LAP results in the mature protein, which functions as a disulfide-linked homodimer. As with all members of the family, TGF beta 3 is highly conserved across species, with mouse and human TGF beta 3 demonstrating 100% sequence homology and cross-species activity.The members of this family can be expressed by most cell types and exert pleiotropic effects, which include the suppression of B- and T-cell effector activity, mediation of tissue healing, and suppression of tumor proliferation. The promotion of CD4+CD25+ T-cell expansion is a newly discovered function of the TGF beta cytokines, and indicates an important role in the protection against autoimmunity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P10600
Assay Range 4.3-1500 pg/mL
Assay Sensitivity 4.3 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias ARVD; ARVD1; FLJ16571; H-TGF-b-3; LAP; Latency-associated peptide; LDS5; prepro-transforming growth factor beta-3; RNHF; TGF beta 3; Tgfb3; TGF-B3; Tgfb-3; TGF-beta3; TGF-beta-3; Transforming growth factor; transforming growth factor beta 3; transforming growth factor beta precursor; transforming growth factor beta-3; Transforming growth factor beta-3 proprotein; transforming growth factor, beta 3; transforming growth factor-beta3ARVD; ARVD1; FLJ16571; H-TGF-b-3; LAP; Latency-associated peptide; LDS5; prepro-transforming growth factor beta-3; RNHF; TGF beta 3; Tgfb3; TGF-B3; Tgfb-3; TGF-beta3; TGF-beta-3; Transforming growth factor; transforming growth factor beta 3; transforming growth factor beta precursor; transforming growth factor beta-3; Transforming growth factor beta-3 proprotein; transforming growth factor, beta 3; transforming growth factor-beta3;
Gene ID (Entrez) 7043
Gene Symbol TGFB3
Interassay CV 12%
Intraassay CV 10%
Kit Contents Pre-coated microwell plate, Standard, Biotinylated Detection Antibody, Streptavidin-HRP, Sample Diluent, Wash Buffer Concentrate, Assay Buffer concentrate, Stop Solution, Substrate Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Shipping Condition Wet Ice
Storage Requirements -20°C
Target TGF beta-3
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.