Learn More
Biomatik Corporation Human TGF Beta Inducible Early Response Gene 1 (TIEG1) ELISA Kit
Supplier:  Biomatik Corporation EKU07610
Encompass_Preferred
Human TGF Beta Inducible Early Response Gene 1 (TIEG1) ELISA Kit for detecting TIEG1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.119ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13118. Gene ID: 7071. Target Synonyms: KLF10; EGRA; TIEG; Krueppel-Like Factor 10; TGFB Inducible Early Growth Response; EGR-alpha; Transforming growth factor-beta-inducible early growth response protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7786