Learn More
ACROBiosystems Human TGF-beta RI 100ug
Catalog No. 502534064
Encompass_Preferred
Human TGF-beta RI Protein Fc Tag (TG1-H5254) is expressed from human 293 cells (HEK293). It contains AA Leu 34 - Leu 126 (Accession # P36897-1).
Catalog No. 50-253-4064 Supplier ACROBiosystems Supplier No. TG1H5254100UG
Human TGF-beta RI Protein Fc Tag (TG1-H5254) is expressed from human 293 cells (HEK293). It contains AA Leu 34 - Leu 126 (Accession # P36897-1).