Learn More
ACROBiosystems Human TGF-beta RI 100ug
Catalog No. 502534062
Encompass_Preferred
Human TGF-beta RI Protein Llama IgG2b Fc Tag (TG1-H5259) is expressed from human 293 cells (HEK293). It contains AA Leu 34 - Leu 126 (Accession # P36897-1).
Catalog No. 50-253-4062 Supplier ACROBiosystems Supplier No. TG1H5259100UG
Human TGF-beta RI Protein Llama IgG2b Fc Tag (TG1-H5259) is expressed from human 293 cells (HEK293). It contains AA Leu 34 - Leu 126 (Accession # P36897-1).