Learn More
ACROBiosystems Human TGF-beta RII 100ug
Catalog No. 502534060
Encompass_Preferred
Human TGF-beta RII Protein Llama IgG2b Fc Tag (TG2-H5253) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Asp 159 (Accession # NP_003233).
Catalog No. 50-253-4060 Supplier ACROBiosystems Supplier No. TG2H5253100UG
Human TGF-beta RII Protein Llama IgG2b Fc Tag (TG2-H5253) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Asp 159 (Accession # NP_003233).