Learn More
Raybiotech Inc Human TGF beta R3 ELISA Kit
Supplier:  Raybiotech Inc ELHTGFBR31
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 40 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair shows no cross-reactivity with recombinant human (rh) TGF-beta RI, RII, RIIb, TGF-beta 1, beta 2, beta 3, recombinant mouse TGF-beta RIII.
Catalog No. 50-194-9772