Learn More
Bon Opus Biosciences Recombinant Human Transforming Growth Factor β-1/TGFB1 1mg
SDP
Supplier:  Bon Opus Biosciences CA591MG
Encompass_Preferred
Recombinant Human Transforming Growth Factor beta 1 is produced by our Mammalian expression system and the target gene encoding Ala279-Ser390 is expressed. Accession# P01137. Formulation: Lyophilized from a 0.2 μm filtered solution of 50mM Glycine 50mM NaCl pH4.0. Bon Opus Cat# CA59
Catalog No. 50-217-2770