Learn More
Sino Biological Human TGFB1 & GARP Protein 1MG (Sino Biological)
Catalog No. 502374200
Encompass_Preferred
A DNA sequence encoding the human TGFB1 (P01137)(Met1-Ser390) was expressed with a polyhistidine tag at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the human GARP (Q14392) (Met1-Asn627) was expressed with a polyhistidine tag at the C-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the TGF Beta 1 & GARP Heterotrimer was purified.
Catalog No. 50-237-4200 Supplier Sino Biological Supplier No. SEKA106025
A DNA sequence encoding the human TGFB1 (P01137)(Met1-Ser390) was expressed with a polyhistidine tag at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the human GARP (Q14392) (Met1-Asn627) was expressed with a polyhistidine tag at the C-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the TGF Beta 1 & GARP Heterotrimer was purified.