Learn More
Sino Biological Anti-TGF beta 1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10804T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human TGF-beta 1/TGFB1 (rh TGF-beta 1/TGFB1; Catalog#10804-H08H; NP_000651.3; Met1-Ser390). TGF-beta 1/TGFB1 specific IgG was purified by Human TGF-beta 1/TGFB1 affinity chromatography.
Catalog No. 50-241-1535