Learn More
Sino Biological Anti-TGF beta 1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10804RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human TGFB1 / TGF-beta1 (rh TGF-beta1; Catalog#10804-H08H; NP_000651.3; Met 1-Ser 390). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1951