Learn More
Sino Biological Human TGFB1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10804RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human TGFB1 / TGF-beta1 (rh TGF-beta1; Catalog#10804-H08H; NP_000651.3; Met 1-Ser 390). TGF-beta1 specific IgG was purified by human TGF-beta1 affinity chromatography.
Catalog No. 50-237-3907