Learn More
Sino Biological Recombinant Human TGF beta 1 Protein (LAP, His Tag)
Supplier:  Sino Biological 10804H08H150
Encompass_Preferred
A DNA sequence encoding the Latency-associated peptide(LAP)of human TGFB1(Met1-Arg278) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-5164