Learn More
BIOHIPPO Human TGFb2 Microsample
SDP
Catalog No. 5000315407
Encompass
Human TGFb2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TGFb2 in Serum plasma tissue homogenates cell culture supernates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 9 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TGFb2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TGFb2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TGFb2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15407 Supplier BIOHIPPO Supplier No. BHE15211435
Human TGFb2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TGFb2 in Serum plasma tissue homogenates cell culture supernates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 9 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TGFb2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TGFb2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TGFb2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color