Learn More
Bon Opus Biosciences Recombinant Human TGFB3 (C-hFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP109050UG
Encompass_Preferred
Recombinant Human TGFB3 (C-hFc) is produced by Human Cells. The target gene encoding Ala301-Ser412 is expressed with a C-hFc tag. Accession# P10600-1. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1090
Catalog No. 50-217-2688