Learn More
BIOHIPPO Human TGFbI Microsample
SDP
Catalog No. 5000315383
Encompass
Human TGFbI Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TGFbI in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 63-40 ng/mL with a sensitivity of 0 237 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TGFbI Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TGFbI Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TGFbI biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15383 Supplier BIOHIPPO Supplier No. BHE15211902
Human TGFbI Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TGFbI in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 63-40 ng/mL with a sensitivity of 0 237 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TGFbI Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TGFbI Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TGFbI biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color