Learn More
Sino Biological Anti-TGF-beta RII/TGFBR2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10358T1620
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human TGFBR2 (rh TGFBR2; NP_003233.4; Met 1-Asp 159). TGFBR2 specific IgG was purified by Human TGFBR2 affinity chromatography.
Catalog No. 50-241-0125