Learn More
ACROBiosystems Human Soluble BCMA ELISA Kit
Catalog No. 502719766
Encompass_Preferred
ClinMax™ ELISA Kit is convenient ready-to-use immunoassay Kit, specifically designed to quantitate human soluble BCMA that is present in complex biological samples, such as human serum, plasma, and cell culture supernates. A comprehensive validation of the ELISA method was performed following the ICH M10 on bioanalytical method validation and the FDA’s bioanalytical method validation guidance for industry. This validation included assessments of linearity, accuracy, precision, dilution linearity, recovery, and the hook effect. For details information, please refer to the DS. ClinMax™ ELISA Kits are manufactured in a GMP-like facility and comply to the ISO 13485 standard, ensuring a high level of quality and reliability.
Catalog No. 50-271-9766 Supplier ACROBiosystems Supplier No. CEAB04596TESTS
ClinMax™ ELISA Kit is convenient ready-to-use immunoassay Kit, specifically designed to quantitate human soluble BCMA that is present in complex biological samples, such as human serum, plasma, and cell culture supernates. A comprehensive validation of the ELISA method was performed following the ICH M10 on bioanalytical method validation and the FDA’s bioanalytical method validation guidance for industry. This validation included assessments of linearity, accuracy, precision, dilution linearity, recovery, and the hook effect. For details information, please refer to the DS. ClinMax™ ELISA Kits are manufactured in a GMP-like facility and comply to the ISO 13485 standard, ensuring a high level of quality and reliability.