Learn More
Raybiotech Inc G-Series Human Th1/Th2 Array 1 Kit.
Supplier:  Raybiotech Inc GSHTH12
Encompass_Preferred
Semi-quantitative measurement of 10 human cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: GM-CSF, IFN-gamma, IL-10, IL-13, IL-2, IL-4, IL-5, IL-6, IL-8 (CXCL8), TNF alpha
Catalog No. 50-195-0429