Learn More
Raybiotech Inc Quantibody Human Th1/Th2 Array 1 Kit.
Supplier:  Raybiotech Inc QAHTH12
Encompass_Preferred
Detects 10 Human Th1 and Th2 Cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: GM-CSF, IFN-gamma, IL-10, IL-13, IL-2, IL-4, IL-5, IL-6, IL-8 (CXCL8), TNF alpha
Catalog No. 50-195-0856