Learn More
Raybiotech Inc C-Series Human Th1/2/17 Antibody Array 1 Kit.
Catalog No. 501948668
Encompass_Preferred
Detects 34 Human Th1, Th2, and Th17 Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: CD30 (TNFRSF8), CD40 Ligand (TNFSF5), CD40 (TNFRSF5), GCSF, GITR (TNFRSF18), GM-CSF, IFN-gamma, IL-1 beta (IL-1 F2), IL-1 R1, IL-1 R2, IL-2, IL-4, IL-5, IL-6, IL-6 R, IL-10, IL-12 p40, IL-12 p70, IL-13, IL-17A, IL-17F, IL-17 RA, IL-21, IL-21 R, IL-22, IL-23, IL-28A (IFN-lambda 2), MIP-3 alpha (CCL20), gp130, TGF beta 1, TGF beta 3, TNF alpha, TNF beta (TNFSF1B), TRANCE (TNFSF11)
Catalog No. 50-194-8668 Supplier Raybiotech Inc Supplier No. AAHTH1718
Detects 34 Human Th1, Th2, and Th17 Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: CD30 (TNFRSF8), CD40 Ligand (TNFSF5), CD40 (TNFRSF5), GCSF, GITR (TNFRSF18), GM-CSF, IFN-gamma, IL-1 beta (IL-1 F2), IL-1 R1, IL-1 R2, IL-2, IL-4, IL-5, IL-6, IL-6 R, IL-10, IL-12 p40, IL-12 p70, IL-13, IL-17A, IL-17F, IL-17 RA, IL-21, IL-21 R, IL-22, IL-23, IL-28A (IFN-lambda 2), MIP-3 alpha (CCL20), gp130, TGF beta 1, TGF beta 3, TNF alpha, TNF beta (TNFSF1B), TRANCE (TNFSF11)