Learn More
Raybiotech Inc G-Series Human Th1/Th2/Th17 Array 1 Kit.
Supplier:  Raybiotech Inc GSHTH1712
Encompass_Preferred
Semi-quantitative measurement of 20 Human TH1-TH2-TH17 cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: GM-CSF, IFN-gamma, IL-1 beta (IL-1 F2), IL-10, IL-12 p70, IL-13, IL-17A, IL-17F, IL-2, IL-21, IL-22, IL-23, IL-28A (IFN-lambda 2), IL-4, IL-5, IL-6, MIP-3 alpha (CCL20), TGF beta 1, TNF alpha, TNF beta (TNFSF1B)
Catalog No. 50-195-0431