Learn More
Sino Biological Anti-Thrombomodulin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 204191T08100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human Thrombomodulin/THBD fragment, and purified by antigen affinity chromatography.
Catalog No. 50-255-3895